4-Amino-7-bromo-pyrrolo[2,1-f][1,2,4]triazine

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 7-Bromopyrrolo[2,1-f][1,2,4]triazin-4-amine

468700010 468700050
468700010.EA 96.6 EUR
468700010. 468700050.
4-Amino-7-bromo-pyrrolo[2,1-f][1,2,4]triazine
4-Amino-7-bromo-pyrrolo[2,1-f][1,2,4]triazine
Formel: C₆H₅BrN₄
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD09033848
CAS-Nummer: 937046-98-5

Bestellen

Spezifikation Testergebnisse

Appearance white to light yellow powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Titration with HClO4 96 to 104 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR