(3R,4R,5R)-3,4-Bis(benzyloxy)-5-((benzyloxy)methyl)dihydrofuran-2(3H)-one
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2,3,5-Tri-O-benzyl-D-ribono-1,4-lactone
468640010
468640050
468640010.EA
36.7
EUR
468640010.
468640050.
(3R,4R,5R)-3,4-Bis(benzyloxy)-5-((benzyloxy)methyl)dihydrofuran-2(3H)-one
(3R,4R,5R)-3,4-Bis(benzyloxy)-5-((benzyloxy)methyl)dihydrofuran-2(3H)-one
Formel:
C₂₆H₂₆O₅ Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD08703966 CAS-Nummer: 55094-52-5 |
Spezifikation Testergebnisse
Appearance | white to light brown powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Melting point | 53°C to 55°C |
Water | ≤0.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...