1-(2-(Azocan-1-yl)ethyl)guanidine sulphate

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Guanethidine sulphate

468620010 468622500
468620010.EA 246 EUR
468620010. 468622500.
1-(2-(Azocan-1-yl)ethyl)guanidine sulphate
1-(2-(Azocan-1-yl)ethyl)guanidine sulphate
Formel: C₁₀H₂₄N₄O₄S
Molekulargewicht: 296.39 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00035403
CAS-Nummer: 645-43-2
EINECS: 211-442-0

Bestellen

Spezifikation Testergebnisse

Appearance white to light beige crystalline powder or crystals
Infrared spectrum Conforms
Titration with HClO4 97.5 to 102.5 %
Loss on drying ≤0.5 % (105°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR