2-Phenylinden

Lieferant: Thermo Fisher Scientific

467770010 467770050
467770010.EA 80.1 EUR
467770010. 467770050.
2-Phenylinden
2-Phenylinden
Formel: C₁₅H₁₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00239514
CAS-Nummer: 4505-48-0

Bestellen

Spezifikation Testergebnisse

Appearance white to light yellow to light brown powder or chunks
Infrared spectrum Conforms
HPLC ≥96.0 %
Melting point 165°C to 169°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR