Estramustine phosphate sodium

Lieferant: Thermo Fisher Scientific
Gefahr

466740010 466742500
466740010.EA 589 EUR
466740010. 466742500.
Estramustine phosphate sodium
Estramustine phosphate sodium
Formel: C₂₃H₃₀Cl₂NNa₂O₆P
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00866320
CAS-Nummer: 52205-73-9
EINECS: 257-735-7
Merck Index: 13,03741

Bestellen

Spezifikation Testergebnisse

Appearance white to almost white powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Water ≤12.5 %
Solubility (50 mg/ml,H2O) Clear colorless to light yellow


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR