4-Chloropyrimidine hydrochloride 97%

Lieferant: Thermo Fisher Scientific
Achtung

457280010 457280050
457280010.EA 89.8 EUR
457280010. 457280050.
4-Chloropyrimidine hydrochloride 97%
4-Chloropyrimidine hydrochloride
Formel: C₄H₃ClN₂·ClH
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD11870807
CAS-Nummer: 179051-78-6

Bestellen

Spezifikation Testergebnisse

Appearance white to brown powder
Infrared spectrum Conforms
Titration with HClO4 96.0 to 104.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR