Brilliant black BN
Lieferant: Thermo Fisher Scientific
199290050
199290250
199290050.EA
20.4
EUR
199290050.
199290250.
Brilliant black BN
Brilliant black BN
Formel:
C₂₈H₁₇N₅O₁₄S₄ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00078980 CAS-Nummer: 2519-30-4 EINECS: 219-746-5 |
Spezifikation Testergebnisse
Appearance | black powder |
Infrared spectrum | Conforms |
UV Lambda max | 567 to 573 nm |
Specific extinction E (1%/1cm) | ≥25000 (15 mg/L in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...