4-[(4-Nitrophenyl)sulfanyl]anilin 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Aminophenyl 4-Nitrophenyl sulphide, 4-((4-Nitrophenyl)thio)aniline, 4-(4-Nitrophenylthio)aniline

104180010 104180050
104180010.EA 34.4 EUR
104180010. 104180050.
4-[(4-Nitrophenyl)sulfanyl]anilin 98%
4-[(4-Nitrophenyl)sulfanyl]anilin
Formel: C₁₂H₁₀N₂O₂S
Molekulargewicht: 246.28 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00007881
CAS-Nummer: 101-59-7
EINECS: 202-957-1

Bestellen

Spezifikation Testergebnisse

Appearance light yellow to brown crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 141°C to 145°C
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR