Bortriiodid ≥95%

Lieferant: Thermo Fisher Scientific

87220.03 87220.06 87220.14
87220.03EA 104 EUR
87220.03 87220.06 87220.14
Bortriiodid ≥95%
Bortriiodid
Formel: BI₃
MDL Number: MFCD00036286
CAS-Nummer: 13517-10-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR