97+%
Lieferant: Thermo Fisher Scientific
Synonyme: 4-Nitrophenyl phosphate disodium salt hexahydrate, Phosphatase substrate, p-Nitrophenyl phosphate disodium salt hexahydrate, Phosphoric acid 4-nitrophenyl ester disodium salt hexahydrate, 4-Nitrophenyl phosphate disodium hexahydrate, pNPP disodium salt hexahydrate, di-Sodium 4-nitrophenyl phosphate hexahydrate
A12310.06 A12310.14 A12310.22
A12310.06EA 58.6 EUR
A12310.06 A12310.14 A12310.22
97+%
di-Natrium-4-nitrophenylphosphat Hexahydrat (pNPP)
Substrate for alkaline and acid phosphatase determinations.
4-Nitrophenyl phosphate disodium salt hexahydrate is a chromogenic substrate used in the determination of acid and alkaline phosphatases.
Formel: C₆H₄NNa₂O₆P·6H₂O
Molekulargewicht: 371.144 g/mol
Schmelzpunkt: >300 °C
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00066288
CAS-Nummer: 333338-18-4
EINECS: 224-246-5