Decyl-β-D-glucopyranosid

Lieferant: BIOMOL RESEARCH LABORATORIES
Achtung

Synonyme: (2R,3R,4S,5S,6R)-2-(Decyloxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, N-Decyl beta-D-glucopyranoside, Decyl β-D-glucopyranoside, Decyl glucopyranoside, DG, Decyl beta-D-glucopyranoside

51458-1 51458-2
BMOL51458-1EA 0 EUR
BMOL51458-1 BMOL51458-2
Decyl-β-D-glucopyranosid
Decyl-β-D-glucopyranosid
Formel: C₁₆H₃₂O₆
Molekulargewicht: 320.43 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00063297
CAS-Nummer: 58846-77-8
EINECS: 261-469-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR