X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid) ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
BCIG, X-GAL, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
Purity: 98+%
CAS No.: 7240-90-6
Molecular Formula: C14H15BrClNO6
Formula Weight: 408.64
Storage Temperature: 2°C to 8°C
Melting Point: ca 236° dec.
Beilstein Reference: 1402009
Optical Rotation: -62° (c=1 in 50% DMF)
MDL No.: MFCD00005666
Formel:
C₁₄H₁₅BrClNO₆ Molekulargewicht: 408.63 g/mol Schmelzpunkt: ∼ 236 °C (Zers.) Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00005666 CAS-Nummer: 7240-90-6 EINECS: 230-640-8 |