Chromotropsäure Dinatriumsalz Dihydrat ACS
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4,5-Dihydroxynaphthalene-2,7-disulphonic acid disodium salt dihydrate, Disodium 4,5-dihydroxynaphthalene-2,7-disulphonate dihydrate, 4,5-Dihydroxynaphthalene-2,7-disulphonic acid, disodium salt dihydrate, 4,5-Dihydroxy-2,7-naphthalenedisulphonic acid disodium salt dihydrate
300600100
300600250
300601000
ACRO300600100EA
33.3
EUR
ACRO300600100
ACRO300600250
ACRO300601000
Chromotropsäure Dinatriumsalz Dihydrat ACS
Chromotropsäure Dinatriumsalz Dihydrat
Formel:
(HO)₂C₁₀H₄(SO₃Na)₂·2H₂O Schmelzpunkt: >300 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00150612 CAS-Nummer: 5808-22-0 EINECS: 204-972-9 Merck Index: 13,02261 |
Spezifikation Testergebnisse
Appearance | Off-white to beige to grey to light brown Powder |
Infrared spectrum | Conforms |
Sensitivity | (to formaldehyde) Passes test |
Clarity of solution | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...