Showa 234X Cut Resistant Gloves, Uncoated
Lieferant: Showa
234X-10/XXL 234X-6/S 234X-7/M 234X-8/L 234X-9/XL
BEST234X-10/XXLEA 98 EUR
BEST234X-10/XXL BEST234X-6/S BEST234X-7/M BEST234X-8/L BEST234X-9/XL
Showa 234X Cut Resistant Gloves, Uncoated
Handschuhe
Designed for use in 'knife hand' applications in food processing.
  • Cut-resistant and abrasion-resistant
  • Flexible and breathable
  • Lightweight and increased dexterity
  • Machine washable
  • Seamless knit
  • Heat-resistant and hydrocarbon-resistant
  • Form-fitting
This flexible cut protection glove is made with uncoated spandex and a HPPE reinforced liner. Cooling properties of HPPE and a breathable design reduce perspiration to keep hands dry, making the 234X ideal for use as a liner glove.
Zertifizierungen: Cat II  EN 388:2016+A1:2018, 1X41D. EN 407:2020, X1XXXX. Food contact approved.
Verpackung: Gloves are packaged 6 pair per case.