iFluor® Ultra 647 succinimidyl ester
Lieferant: AAT BIOQUEST
iFluor® Ultra 647 is an exceptionally bright far-red fluorescent dye excitable by 594 nm or 633 nm laser lines, with a peak emission at 670 nm.
- Stable for 12 months
Fluorescent dye-conjugated antibodies provide a tool for identifying proteins in many applications including fluorescent cell imaging, flow cytometry, western blotting, immunohistochemistry and more.
Bitte beachten For Research Use Only