Anti-Aurora A Mouse Monoclonal Antibody [clone: 4A7]

Lieferant: ANTIBODIES.COM
A85401-100 A85401-50
ANTIA85401-100EA 520 EUR
ANTIA85401-100 ANTIA85401-50
Anti-Aurora A Mouse Monoclonal Antibody [clone: 4A7]
Antikörper
Mouse monoclonal (4A7) antibody to Aurora A + B + C Kinase for WB, ICC/IF and IHC with samples derived from Human, Horse, Bovine, Porcine, Rat and Mouse.

  • Validated applications: WB, ICC/IF, IHC
  • Recommended dilutions: WB: 1:1000, ICC/IF: 1:1000 to 1:2000, IHC: 1:1000 to 1:200

Type: Primary
Antigen: Aurora A
Clonality: Monoclonal
Clone: 4A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Horse;Bovine;Porcine;Rat;Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR