Anti-Aurora A Mouse Monoclonal Antibody [clone: 5A12]
Lieferant: ANTIBODIES.COM
A85399-100 A85399-50
ANTIA85399-100EA 520 EUR
ANTIA85399-100 ANTIA85399-50
Anti-Aurora A Mouse Monoclonal Antibody [clone: 5A12]
Antikörper
Mouse monoclonal (5A12) antibody to Aurora A + B Kinase for WB, ICC/IF and IHC with samples derived from Human, Horse, Rat, Mouse and Canine.
  • Validated applications: WB, ICC/IF, IHC
  • Recommended dilutions: WB: 1:1000 to 1;2000, ICC/IF: 1:1000 to 1;2000, IHC: 1:1000 to 1:200
Type: Primary
Antigen: Aurora A
Clonality: Monoclonal
Clone: 5A12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Horse;Rat;Mouse;Dog