Eppendorf® Xplorer® Plus Move It® Electronic Multi-Channel Pipettes With Adjustable Tip Spacing
Lieferant: EPPENDORF
4861000.781 4861000.782 4861000.794 4861000.795 4861000.816 4861000.817 4861000.818 4861000.833 4861000.834 4861000.835
613-6988EA 2155 EUR
613-6988 613-6989 613-6992 613-6993 613-6891 613-6892 613-6893 613-6894 613-6895 613-6896
Eppendorf® Xplorer® Plus Move It® Electronic Multi-Channel Pipettes With Adjustable Tip Spacing
Pipetten Mikropipetten
Eppendorf® Xplorer® Plus Move It® Electronic multi-channel pipettes with adjustable cones allow easy vessel format changes, cutting up to 50% of your time compared to single-channel pipettes. With several pipetting modes for enhanced productivity and offered in 4, 6, 8 or 12 channels.
  • Air-cushion pipettes with variable volume and adjustable tip spacing are available in 4, 6, 8 and 12-channel
  • Choose from Mechanical (Research™ plus) or Electronic (Xplorer™ plus) adjustable tip spacing multichannel pipettes
  • For Plates (12, 24, 48, 96, and 384-well), Tubes (1,5 and 2 ml), and Agarose gels
  • Tip spacing is freely selectable from 4,5 to 33 mm, depending on the pipette variant
  • Spacing control for quick switches backwards and forwards between source and destination formats
  • Manual adjustment knob for smooth, vibration-free format switches and drip-free liquid transfers
  • 360° rotatable pipette head for comfortable readability of display and fast parameter identification
  • Tubeless system is safe and reliable and optimally balanced pipette reduces hand fatigue for stress-free operation
Air-cushion pipetting system and autoclavable for user and sample safety (Research™ plus fully, and Xplorer™ plus lower part only).
Applications: Pipetting of aqueous liquids