Anti-Dextran Mouse Monoclonal Antibody [clone: DX1] (FITC)

Lieferant: STEMCELL Technologies
60026FI 60026FI.1
STMC60026FIEA 335 EUR
STMC60026FI STMC60026FI.1
Anti-Dextran Mouse Monoclonal Antibody [clone: DX1] (FITC)
Antikörper
Mouse monoclonal IgG1 antibody against dextran, FITC-conjugated.

The DX1 antibody reacts with dextran, a natural polysaccharide consisting of linear chains of alpha-1,6-linked D-glucopyranose residues with short side chains (mostly 1 to 2 glucose units) 1,3- or 1,4-linked to the backbone of the biopolymer. Dextran has several uses as an additive in food, in lacquers, as a plasma volume expander, and as a coating for particles used in bioimaging or cell separation applications. The DX1 antibody was raised against isomaltotetraose (four sugar units in length) but binds better to longer polymer chains. It is also known to bind to amino-dextran, though with lower affinity. This antibody can be used to detect cells labelled with dextran-coated nanoparticles during StemSep™ and EasySep™ cell separation procedures or for other applications that require the detection of dextran. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ human T cell enrichment kit (Catalog #19051) and EasySep™ human CD4+ T cell enrichment kit (Catalog #19052).

Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type: Anti-Dextran Mouse Monoclonal Antibody [clone: DX1] (FITC)
Antigen: Dextran
Clonality: Monoclonal
Clone: DX1
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR