Anti-CD34 Mouse Monoclonal Antibody [clone: 581] (FITC)

Lieferant: STEMCELL Technologies
60013FI 60013FI.1
STMC60013FIEA 473 EUR
STMC60013FI STMC60013FI.1
Anti-CD34 Mouse Monoclonal Antibody [clone: 581] (FITC)
Antikörper
Mouse monoclonal IgG1 antibody against human, chimpanzee, horse CD38, PE­conjugated.

  • B cells
  • Hematopoietic stem
  • Progenitor cells
  • Plasma

The HIT2 antibody reacts with CD38, an ~45 kDa type II transmembrane glycoprotein expressed by many cell types, especially leukocytes. CD38 is an ectoenzyme (ADP-ribosyl hydrolase) with both cyclase and hydrolase activities, whereby it mediates lymphocyte activation, adhesion and metabolism. CD31 is a known ligand for the receptor. In humans, CD38 is expressed by the majority of hematopoietic cells at levels which vary according to the differentiation and activation status of the cells. It is highly expressed on activated T and B cells and plasma cells, and is also found at high levels in several tissues, including brain, kidney, liver, ovary, pancreas, placenta, testis, and some types of cancerous tissue. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ human CD138 positive selection kit II (Catalog #1877) and EasySep™ human whole blood and Bone Marrow CD138 positive selection kit II (Catalog #17887).

Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type: Anti-CD34 Mouse Monoclonal Antibody [clone: 581] (FITC)
Antigen: CD34
Clonality: Monoclonal
Clone: 581
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR