Methyl-4-chlor-3-nitrobenzoat ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Chloro-3-nitrobenzoic acid methyl ester, Methyl 4-chloro-3-nitrobenzate

L07407.06 L07407.14
L07407.06EA 49.2 EUR
L07407.06 L07407.14
Methyl-4-chlor-3-nitrobenzoat ≥98%
Methyl-4-chlor-3-nitrobenzoat

Purity: 98% CAS No.: 14719-83-6 Molecular Formula: C8H6ClNO4 Formula Weight: 215.59 Melting Point: 78-82° Beilstein Reference: 1643729

Formel: C₈H₆ClNO₄
Molekulargewicht: 215.59 g/mol
Schmelzpunkt: 78…82 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00016989
CAS-Nummer: 14719-83-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR