Methyl-2-chlor-5-nitrobenzoat

Lieferant: Thermo Scientific

Synonyme: 2-Chloro-5-nitrobenzoic acid methyl ester, Methyl 2-chloro-5-nitrobenzenecarboxylate

RJF01607.1 RJF01607.10 RJF01607.50
MAYBRJF01607.1EA 0 EUR
MAYBRJF01607.1 MAYBRJF01607.10 MAYBRJF01607.50
Methyl-2-chlor-5-nitrobenzoat
Methyl-2-chlor-5-nitrobenzoat
Formel: C₈H₆ClNO₄
Molekulargewicht: 215.59 g/mol
MDL Number: MFCD00184587
CAS-Nummer: 6307-82-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR