Gram Jodlösung, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme: Lugol's iodine, Iodine and potassium iodide, Potassium triiodide
M-Clarity™ Qualitätsstufe = MQ500
HT90232-1L HT902-8FOZ
HT90232-1LEA 165 EUR
HT90232-1L HT902-8FOZ
Gram Jodlösung, Sigma-Aldrich®
Gram Jodlösung
Used as a kit component in Gram Stain Procedure (Sigma Procedure HT90). Iodine, 0.33%, and potassium iodide, 0.66%.
Formel: I₃K
Molekulargewicht: 419.812 g/mol
Lagerungstemperatur: Room Temperature
MDL Number: MFCD00134128
CAS-Nummer: 12298-68-9
IVDLogo
Bestellen
Spezifikation Testergebnisse
Appearance (Colour) Orange-Brown
Appearance (Form) Solution
Appearance (Turbidity)
Essentially free of particles/precipitate
Clear
Performance Conforms
Gram positive organisms are purple-black in both bacteriology smears and tissue sections; Gram negative organisms are red in both bacteriology smears and tissue sections
Expiration Date Period 24 months