Pyridiniumchlorochromat ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A11752.14
A11752.22
A11752.36
A11752.PG
A11752.14EA
26.3
EUR
A11752.14
A11752.22
A11752.36
A11752.PG
Pyridiniumchlorochromat ≥98%
Pyridinium chlorochromate
Purity: 98%
CAS No.: 26299-14-9
Molecular Formula: C5H6ClCrNO3
Formula Weight: 215.56
Melting Point: ca 206° dec.
Merck Reference: 14,7974
MDL No.: MFCD00013106
Formel:
C₅H₆ClCrNO₃ Schmelzpunkt: ∼ 206 °C (dec.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00013106 CAS-Nummer: 26299-14-9 EINECS: 247-595-5 UN-Nummer: 1479 ADR: 5.1,II Merck Index: 14,7974 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...