Phenolrot Natriumsalz, wasserlöslich, rein Indikator
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, sodium salt, Phenol red water soluble, Sodium hydrogen 4,4'-(3H-2,1-benzoxathiol-3-ylidene)bisphenolate S,S-dioxide
151440250
151440050
151441000
ACRO151440250EA
79.2
EUR
ACRO151440250
ACRO151440050
ACRO151441000
Phenolrot Natriumsalz, wasserlöslich, rein Indikator
Phenolrot Natriumsalz
Formel:
C₁₉H₁₃NaO₅S Molekulargewicht: 376.35 g/mol Schmelzpunkt: 285 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00066901 CAS-Nummer: 34487-61-1 EINECS: 252-057-8 Merck Index: 12,07397 |
Spezifikation Testergebnisse
Appearance | Dark red to brown Powder |
Infrared spectrum | Conforms |
Solubility | (0.1 % in water) Clear |
Specific extinction E (1%/1cm) | ≥1600 (558 nm, in buffer) |
Visual transition interval | from pH 6.8 (yellow) to pH 8.2 (red) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...