F(ab') Anti-IgG (H+L) Goat Polyclonal Antibody (CF® 660C)

Lieferant: Biotium
20917-50UL 20917-250UL
20917-50ULEA 0 EUR
20917-50UL 20917-250UL
F(ab') Anti-IgG (H+L) Goat Polyclonal Antibody (CF® 660C)
Antikörper
This is a goat anti-rabbit IgG (H L) F(ab’)2 fragment secondary antibody labeled with superior CF® Dyes and other labels.

  • Suitable for western, immunofluorescence, and immunohistology in FFPE tissues

Liquid format: Store at −20 °C, protected from light. Product is stable for at least 6 months from date of receipt when stored as recommended. Liquid format antibodies contain 50% glycerol and will not freeze at −20 °C.
Lyophilised format: Store at −20 °C, protected from light. Product is stable for at least 6 months from date of receipt when stored as recommended.

Reconstitute antibodies in water using the indicated volumes below:
CF® Dye and biotin conjugates: add 0,5 ml dH₂O.
HRP or DNP conjugates: add 1 ml dH₂O.
Add the indicated volume of water directly to the vial containing the lyophilised antibody and mix gently to dissolve. Store reconstituted antibody at −20 °C and protect from light. Aliquot to avoid repeated freeze/thaw cycles. Alternatively, an equal volume of glycerol can be mixed with the reconstituted antibody so that it will remain liquid at −20 °C.

Optional: A preservative such as 0,05% sodium azide (final concentration) can be added to CF® Dye and biotin conjugates. Do not add sodium azide to HRP conjugates. Note: Storage of the antibody for more than a day at final working dilution is not recommended.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF660C
Epitope:
Host: Goat
Isotype: IgG (H+L)
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR