Rhodamin 6G, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
9-(2-(Ethoxycarbonyl)phenyl)-3,6-bis(ethylamino)-2,7-dimethylxanthylium chloride, 9-[2-(Ethoxycarbonyl)phenyl]-3,6-bis(ethylamino)-2,7-dimethylxanthylium chloride
252433-250MG
252433-1G
SIAL252433-250MGEA
56
EUR
SIAL252433-250MG
SIAL252433-1G
Rhodamin 6G, Sigma-Aldrich®
Rhodamin 6G
Formel:
C₂₈H₃₁ClN₂O₃ Molekulargewicht: 479.02 g/mol Siedepunkt: ∼200 °C (1013 hPa) |
MDL Number:
MFCD00012665 CAS-Nummer: 989-38-8 EINECS: 213-584-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...