Bromkresolpurpur Natriumsalz, orangenes Pulver

Lieferant: MP Biomedicals
Achtung

Synonyme: Sodium α-(3-bromo-5-methyl-4-oxo-2,5-cyclohexadienylidene)-α-(3-bromo-5-methyl-4-hydroxyphenyl)toluenesulphonate, Bromocresol purple water soluble, Sodium 2-bromo-4-(3-(3-bromo-4-hydroxy-5-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-6-methylphenolate

0215051825 0215051805
ICNA0215051825EA 75.3 EUR
ICNA0215051825 ICNA0215051805
Bromkresolpurpur Natriumsalz, orangenes Pulver
Bromkresolpurpur Natriumsalz

Bromocresol purple sodium salt is typically used as a pH indicator (5,2 - 6,8 : yellow to blue-purple). Used in spectrophotometric determination of serum-albumin.


Soluble in water (80 mg/ml), ethanol (20 mg/ml) or methyl cellosolve (90 mg/ml).

Formel: C₂₁H₁₅Br₂NaO₅S
Molekulargewicht: 562.21 g/mol
Schmelzpunkt: 255 (Zers.)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00148896
CAS-Nummer: 62625-30-3
EINECS: 263-655-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR