Bromthymolblau Natriumsalz

Lieferant: Thermo Fisher Scientific

Synonyme: Sodium α-(3-bromo-5-isopropyl-4-oxo-2-methyl-2,5-cyclohexadienylidene)-2-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)toluenesulphonate, Bromothymol blue water soluble, Sodium 2-bromo-4-(3-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-6-isopropyl-3-methylphenolate

A17913.09 A17913.18
A17913.09EA 43.7 EUR
A17913.09 A17913.18
Bromthymolblau Natriumsalz
Bromthymolblau Natriumsalz

CAS No.: 34722-90-2 Molecular Formula: C27H27Br2NaO5S Formula Weight: 646.38 Melting Point: 200-202° MDL No.: MFCD00077263

Formel: C₂₇H₂₇Br₂NaO₅S
Molekulargewicht: 646.37 g/mol
Schmelzpunkt: 200…202 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00077263
CAS-Nummer: 34722-90-2
EINECS: 252-169-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR