EDTA Eisen(III)mononatriumsalz, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Ethylenedinitrilotetra-acetic acid iron (III) sodium salt, Edetic acid iron (III) monosodium salt, Edetic acid iron (III) sodium salt, Ethylenediaminetetra-acetic acid iron (III) sodium salt, Iron (III) sodium ethylenediaminetetraacetate, Ethylenedinitrilotetra-acetic acid iron (III) monosodium salt, EDTA iron sodium, Sodium ferric ethylenediaminetetraacetate, Iron (III) monosodium ethylenediaminetetraacetate, Sodium feredetate, Ethylenediaminetetra-acetic acid iron (III) monosodium salt

M-Clarity™ quality level = MQ200

EDFS-100G EDFS-1KG EDFS-250G
SIALEDFS-100GEA 116 EUR
SIALEDFS-100G SIALEDFS-1KG SIALEDFS-250G
EDTA Eisen(III)mononatriumsalz, Sigma-Aldrich®
EDTA Eisen(III)mononatriumsalz
Formel: C₁₀H₁₂FeN₂NaO₈
Schmelzpunkt: 80 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00078215
CAS-Nummer: 15708-41-5
EINECS: 239-802-2
Merck Index: 12,04076

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR