Capsazepine ≥98%
Lieferant: ENZO LIFE SCIENCES
Synonyme:
N-(4-Chlorophenethyl)-7,8-dihydroxy-4,5-dihydro-1H-benzo[c]azepine-2(3H)-carbothioamide
Capsaicin antagonist
Analog of capsaicin (Prod. No. BML-EI125-0200) that acts as a capsaicin antagonist. Inhibits resiniferatoxin and capsaicin-induced contractions of guinea pig trachea. Inhibits C-fiber-smooth muscle contractions induced by capsaicin, bradykinin and neurokinin A. Increases intracellular Ca²⁺ levels.
Synthetic.
Formel:
C₁₉H₂₁ClN₂O₂S |
MDL Number:
MFCD00153778 CAS-Nummer: 138977-28-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...