Vancomycin Hydrochlorid (aus Streptomyces lysosup)

Lieferant: ENZO LIFE SCIENCES

Synonyme: (3S,6R,7R,22R,23S,26S,36R,38aR)-3-(2-Amino-2-oxoethyl)-44-[[2-O-(3-amino-2,3,6-trideoxy-3-C-methyl-α-L-lyxo-hexopyranosyl)-β-D-glucopyranosyl]oxy]-10,19-dichloro-2,3,4,5,6,7,23,24,25,26,36,37,38,38a-tetradecahydro-7,22,28,30,32-pentahydroxy-6-[[(2R)-4-methyl-2-(methylamino)-1-oxopentyl]amino]-2,5,24,38,39-pentaoxo-22H-8,11:18,21-Dietheno-23,36-(iminomethano)-13,16:31,35-dimetheno-1H,16H-[1,6,9]oxadiazacyclohexadecino[4,5-m][10,2,16]benzoxadiazacyclotetracosine-26-carboxylic acid xhydrochloride, Vancomycin supplement

ALX380279G001 ALX380279M250
ENZOALX380279G001EA 286 EUR
ENZOALX380279G001 ENZOALX380279M250
Vancomycin Hydrochlorid (aus Streptomyces lysosup)
Vancomycin Hydrochlorid

Glycopeptide antibiotic. Interferes with cell wall synthesis. Effective against Gram-positive bacteria.


White to light brown powder


Soluble in water (50 mg/ml)


Verpackung: Shipping temperature: Ambient

Formel: C₆₆H₇₅Cl₂N₉O₂₄·HCl
Molekulargewicht: 1485.71 g/mol
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD03613611
CAS-Nummer: 1404-93-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR