Thiostrepton
Lieferant: ENZO LIFE SCIENCES
Synonyme:
N-[(7R,8S)-2-Carboxy-7,8-dihydro-8-hydroxy-4-[(1S)-1-hydroxyethyl]-7-quinolinyl]-L-isoleucyl-L-alanyl-2,3-didehydroalanyl-L-alanyl-2-[(4aR,11S,14Z,18S,21S,28S,32aS)-4a-amino-21-[(1S,2R)-1,2-dihydroxy-1-methylpropyl]-14-ethylidene-3,4,4a,9,10,11,12,13,14,18,19,20,21,27,28,32a-hexadecahydro-11,28-bis[(1R)-1-hydroxyethyl]-9,12,19,26-tetraoxo-17H,26H-8,5:18,15:25,22:32,29-tetranitrilo-5H,15H-pyrido[3,2-m][1,11,17,24,4,7,20,27]tetrathiatetraazacyclotriacontin-2-yl]-4-thiazolecarbonyl-2,3-didehydroalanyl-2,3-didehydro-alaninamide,(1→528)-lactone
Elongation factor G inhibitor
Thiazole-containing peptide antibiotic. Inhibits protein synthesis by preventing binding of GTP to the 50S ribosomal subunit. Inhibits binding of elongation factor G (EF-G) and the dissociation of EF-G from the ribosome.
Isolated from Streptomyces azureus.
Formel:
C₇₂H₈₅N₁₉O₁₈S₅ Molekulargewicht: 1664.887 g/mol |
MDL Number:
MFCD00135828 CAS-Nummer: 1393-48-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...