Cytochalasin A

Lieferant: ENZO LIFE SCIENCES

ALX380057M001 ALX380057M005 ALX380057M010
ENZOALX380057M001EA 102 EUR
ENZOALX380057M001 ENZOALX380057M005 ENZOALX380057M010
Cytochalasin A
Cytochalasin A
Formel: C₂₉H₃₅NO₅
Schmelzpunkt: 182…185 °C
MDL Number: MFCD00005935
CAS-Nummer: 14110-64-6
EINECS: 237-964-9
UN-Nummer: 1544
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR