Genistein

Lieferant: Enzo Life Sciences

ALX350006G001 ALX350006M010 ALX350006M025 ALX350006M050 ALX350006M100
ENZOALX350006G001EA 1010 EUR
ENZOALX350006G001 ENZOALX350006M010 ENZOALX350006M025 ENZOALX350006M050 ENZOALX350006M100
Genistein
Genistein
Formel: C₁₅H₁₀O₅
Schmelzpunkt: 297…298 °C
MDL Number: MFCD00016952
CAS-Nummer: 446-72-0
EINECS: 207-174-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR