N-Boc-L-aspartic acid 4-methyl ester fluoromethyl ketone ≥95% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme: Boc-Asp(OMe)-fluoromethyl ketone, Methyl (S)-3-((tert-butoxycarbonyl)amino)-5-fluoro-4-oxopentanoate
ALX260071M001 ALX260071M005
ENZOALX260071M001EA 438 EUR
ENZOALX260071M001 ENZOALX260071M005
N-Boc-L-aspartic acid 4-methyl ester fluoromethyl ketone ≥95% (durch TLC)
N-Boc-L-aspartic acid 4-methyl ester fluoromethyl ketone
Irreversible, cell permeable broad-spectrum caspase inhibitor.
  • Caspase inhibitor
White to off-white powder.
Soluble in DMSO or dimethyl formamide.
Formel: C₁₁H₁₈FNO₅
Molekulargewicht: 263.265 g/mol
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD03453073
CAS-Nummer: 187389-53-3