B581 ≥96%

Lieferant: ENZO LIFE SCIENCES

Synonyme: H-Cys-psi(CH₂NH)Val-psi(CH₂NH)Phe-Met-OH

BMLG2350001 BMLG2350005
ENZOBMLG2350001EA 202 EUR
ENZOBMLG2350001 ENZOBMLG2350005
B581 ≥96%
B581

Farnesyltransferase inhibitor


A potent and selective, cell-permeable farnesyl transferase inhibitor IC50=21 nM, 30-fold selective over geranylgeranyl transferase.

Formel: C₂₂H₃₈N₄O₃S₂
MDL Number: MFCD18910841
CAS-Nummer: 149759-96-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5cef061310b7dac5bbeccb5ced37b03b8f16c75&vid=f80710c6888ef5dc094df34a6e4c4a8cb49f5582&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR