Ketoconazol ≥99%

Lieferant: ENZO LIFE SCIENCES
Gefahr

Synonyme: (+/-)-cis-1-Acetyl-4-[4-[[2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazine, cis-1-Acetyl-4-[4-[[2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazine

BMLEI1070050 BMLEI1070500
ENZOBMLEI1070050EA 71.5 EUR
ENZOBMLEI1070050 ENZOBMLEI1070500
Ketoconazol ≥99%
Ketoconazole

Cytochrome P450 inhibitor


Inhibits cytochrome P-450-dependent steps in the biosynthesis of steroid hormones in vivo. Antimetastatic and antineoplastic activity. Antipsoriatic activity. Orally active 5-lipoxygenase and thromboxane synthase inhibitor.


Bitte beachten Research use only.

Formel: C₂₆H₂₈Cl₂N₄O₄
Molekulargewicht: 531.43 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00058579
CAS-Nummer: 65277-42-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ad3e389cf9656c2221708da9987956e24239f05&vid=6e021355e3974cdfbd7403fdbc6c8e89dc31f5d7&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR