2-Brom-5-pyridinboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Bromo-3-pyridinylboronic acid, 2-Bromopyridine-5-boronic acid, (6-Bromopyridin-3-yl)boronic acid, 2-Bromo-5-pyridinylboronic acid, 6-Bromopyridine-3-boronic acid

M-Clarity™ quality level = MQ100

666556-1G 666556-5G
SIAL666556-1GEA 41.8 EUR
SIAL666556-1G SIAL666556-5G
2-Brom-5-pyridinboronsäure, Sigma-Aldrich®
2-Brom-5-pyridinboronsäure
Formel: C₅H₅BBrNO₂
MDL Number: MFCD03411558
CAS-Nummer: 223463-14-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR