DL-α-Phenylalanin 99%
Lieferant: Thermo Fisher Scientific
Synonyme:
H-DL-Phe-OH, DL-Phenylalanine, 3-Phenyl-DL-alanine
164570250
164571000
164575000
ACRO164570250EA
29.7
EUR
ACRO164570250
ACRO164571000
ACRO164575000
DL-α-Phenylalanin 99%
DL-α-Phenylalanin
Formel:
C₉H₁₁NO₂ Siedepunkt: 313 °C (1013 hPa) Schmelzpunkt: 266 °C (dec.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00064225 CAS-Nummer: 150-30-1 EINECS: 205-756-7 Merck Index: 13,07355 |
Spezifikation Testergebnisse
Appearance | White Powder or crystals |
Infrared spectrum | Conforms |
Loss on drying | ≤0.3 % |
Sulfated ash | ≤0.3 % |
Titration with HClO4 | 98.5 to 101.5 % (on dry substance) |
Arsenic (As) | ≤3 ppm |
Lead (Pb) | ≤10 ppm |
Total nitrogen | ≥9.0 % (on dry substance) |
Amino nitrogen | ≥3.0 % (on dry substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...