2-Fluor-6-iodbenzonitril
Lieferant: Thermo Scientific
CD07951.1
CD07951.10
MAYBCD07951.1EA
0
EUR
MAYBCD07951.1
MAYBCD07951.10
2-Fluor-6-iodbenzonitril
2-Fluor-6-iodbenzonitril
Formel:
C₇H₃FIN Siedepunkt: 281 °C (1013 hPa) Schmelzpunkt: 48,5…50.5 °C |
MDL Number:
MFCD00015478 CAS-Nummer: 79544-29-9 UN-Nummer: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...