(4-Hydroxyphenyl)acetonitril, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-Hydroxybenzyl cyanide, 2-(4-Hydroxyphenyl)acetonitrile
M-Clarity™ quality level = MQ100
H21101-25G
H21101-5G
SIALH21101-25GEA
34.3
EUR
SIALH21101-25G
SIALH21101-5G
(4-Hydroxyphenyl)acetonitril, Sigma-Aldrich®
(4-Hydroxyphenyl)acetonitril
Formel:
C₈H₇NO Siedepunkt: 330 °C (1013 hPa) Schmelzpunkt: 68…71 °C |
MDL Number:
MFCD00002383 CAS-Nummer: 14191-95-8 EINECS: 238-046-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...