D-α-Phenylalanin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: D-Phenylalanine, H-D-Phe-OH, 3-Phenyl-D-alanine, 3-Phenyl-D(+)-alanine

M-Clarity™ quality level = MQ300

P1751-100G P1751-25G P1751-5G
SIALP1751-100GEA 582 EUR
SIALP1751-100G SIALP1751-25G SIALP1751-5G
D-α-Phenylalanin, Sigma-Aldrich®
D-α-Phenylalanin
Formel: C₉H₁₁NO₂
Siedepunkt: 295 °C (1013 hPa)
MDL Number: MFCD00004270
CAS-Nummer: 673-06-3
EINECS: 211-603-5
Merck Index: 12,07425

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR