Kieselgel, Davisil® Grade 62 pore size 150 Å, 35-60 mesh, hochrein, Supelco®
Lieferant: Merck
236845-100G
236845-1KG
236845-100GEA
104
EUR
236845-100G
236845-1KG
Kieselgel, Davisil® Grade 62 pore size 150 Å, 35-60 mesh, hochrein, Supelco®
Silica gel
Formel:
SiO₂ Molekulargewicht: 60.08 g/mol Siedepunkt: 2230 °C Schmelzpunkt: > 1600 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00011232 CAS-Nummer: 63231-67-4 EINECS: 231-545-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...