Bortribromid 1 mol/l in Dichlormethan, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ200
211222-100ML
211222-800ML
SIAL211222-100MLEA
150
EUR
SIAL211222-100ML
SIAL211222-800ML
Bortribromid 1 mol/l in Dichlormethan, Sigma-Aldrich®
Bortribromid
Formel:
BBr₃ |
MDL Number:
MFCD00011312 CAS-Nummer: 10294-33-4 EINECS: 233-657-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...