Ethyl-6-bromhexanoat
Lieferant: Thermo Scientific
Synonyme:
6-Bromocaproic acid ethyl ester, 6-Bromohexanoic acid ethyl ester, Ethyl 6-bromohexanoate
SB01144.1
SB01144.10
SB01144.50
MAYBSB01144.1EA
0
EUR
MAYBSB01144.1
MAYBSB01144.10
MAYBSB01144.50
Ethyl-6-bromhexanoat
Ethyl-6-bromhexanoat
Formel:
BrH₂C(CH₂)₄COOCH₂CH₃ Siedepunkt: 69…72 °C (0,1 mmHg) Schmelzpunkt: 33 °C Dichte: 1,2592 g/cm³ (20 °C) |
MDL Number:
MFCD00000270 CAS-Nummer: 25542-62-5 EINECS: 247-085-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...