δ-Valerolacton, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Pentan-5-olide, Tetrahydro-2H-pyran-2-one, 5-Hydroxypentanoic acid lactone, δ-Valerolactone
M-Clarity™ quality level = MQ100
389579-100ML
389579-25ML
SIAL389579-100MLEA
143
EUR
SIAL389579-100ML
SIAL389579-25ML
δ-Valerolacton, Sigma-Aldrich®
δ-Valerolacton
Formel:
C₅H₈O₂ Siedepunkt: 228…230 °C (1013 hPa) Schmelzpunkt: –10 °C Dichte: 1.0988 g/cm³ (20 °C) |
MDL Number:
MFCD00006645 CAS-Nummer: 542-28-9 EINECS: 208-807-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...