4-Tolylboronsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Dihydroxy(4-methylphenyl)borane, p-Tolylboronic acid, 4-Methylphenylboronic acid, (4-Methylphenyl)metaboric acid, 4-Methylbenzeneboronic acid
M-Clarity™ quality level = MQ100
393622-10G
393622-1G
SIAL393622-10GEA
245
EUR
SIAL393622-10G
SIAL393622-1G
4-Tolylboronsäure, Sigma-Aldrich®
4-Tolylboronsäure
Formel:
C₇H₉BO₂ Schmelzpunkt: 260 °C |
MDL Number:
MFCD00039138 CAS-Nummer: 5720-05-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...