4-Tolylboronsäure 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Dihydroxy(4-methylphenyl)borane, p-Tolylboronic acid, 4-Methylphenylboronic acid, (4-Methylphenyl)metaboric acid, 4-Methylbenzeneboronic acid
344430010
344430050
AC34443.10
344430250
ACRO344430010EA
28.3
EUR
ACRO344430010
ACRO344430050
MAYBAC34443.10
ACRO344430250
4-Tolylboronsäure 97%
4-Tolylboronsäure
Formel:
C₇H₉BO₂ Schmelzpunkt: 256…263 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00039138 CAS-Nummer: 5720-05-8 |
Spezifikation Testergebnisse
Appearance | White to cream to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 256 to 263 °C |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...