4-Tolylboronsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Dihydroxy(4-methylphenyl)borane, p-Tolylboronic acid, 4-Methylphenylboronic acid, (4-Methylphenyl)metaboric acid, 4-Methylbenzeneboronic acid

A13347.03 A13347.06 A13347.14
A13347.03EA 43.7 EUR
A13347.03 A13347.06 A13347.14
4-Tolylboronsäure ≥98%
4-Tolylboronsäure

Purity: 98% CAS No.: 5720-05-8 Molecular Formula: C7H9BO2 Formula Weight: 135.96 Melting Point: 246-254° Beilstein Reference: 2935970

Formel: C₇H₉BO₂
Schmelzpunkt: 246…254 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00039138
CAS-Nummer: 5720-05-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR